missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-PlGF, Clone: PLGF94, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen PlGF
Clone PLGF94
Concentration 1mg/mL
Immunogen Recombinant human PLGF protein
Gene Accession No. P49763
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23471001 Novus Biologicals Canada
NBP23471001MG
0.1mg Each for $384.00
NBP23471002 Novus Biologicals Canada
NBP23471002MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

PlGF
1mg/mL
P49763
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
PLGF94
Recombinant human PLGF protein
5228
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Functional Assay