Please login to your online account to display your discounted pricing

Mouse anti-PlGF, Clone: PLGF94, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23471002MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23471002

  • / Each
 

Description & Specifications

Specifications

Antigen PlGF
Applications ELISA
Applications Functional Assay
Immunogen Recombinant human PLGF protein
Clone PLGF94
Conjugate Unlabeled
Host Species Mouse
Format Purified
Purification Method Purified
Regulatory Status RUO
Quantity 0.2mg
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Accession No. P49763
Concentration 1mg/mL
Cross Reactivity Human
Isotype IgG1, κ
Reconstitution Protein A purified
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Gene ID (Entrez) 5228