Please login to your online account to display your discounted pricing

Mouse anti-PlGF, Clone: PLGF94, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23471001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23471001

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mg
Antigen PlGF
Applications ELISA
Applications Functional Assay
Host Species Mouse
Isotype IgG1, κ
Conjugate Unlabeled
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Immunogen Recombinant human PLGF protein
Gene ID (Entrez) 5228
Format Purified
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Concentration 1mg/mL
Gene Accession No. P49763
Clone PLGF94
Purification Method Purified
Reconstitution Protein A purified
Cross Reactivity Human