Please login to your online account to display your discounted pricing

Mouse anti-Plasma Cell Marker, Clone: SPM310, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Plasma Cell Marker
Clone SPM310
Concentration 0.2mg/mL
Immunogen Pancreatic cancer-related mucin
Formulation PBS with 0.05% BSA.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23435901

 
novus biologicals canada
NBP23435901ML
0.1mL Each for $139.00

NBP23435905

 
novus biologicals canada
NBP23435905ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Plasma Cell Marker
Clone SPM310
Concentration 0.2mg/mL
Immunogen Pancreatic cancer-related mucin
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2a
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry (Paraffin)