Please login to your online account to display your discounted pricing

Mouse anti-Plasma Cell Marker, Clone: SPM310, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Plasma Cell Marker
Clone SPM310
Concentration 1mg/mL
Immunogen Pancreatic cancer-related mucin
Formulation 10mM PBS. Does not contain BSA. Azide Free.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23440701

 
novus biologicals canada
NBP23440701MG
0.1mg Each for $384.00

NBP23440702

 
novus biologicals canada
NBP23440702MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Plasma Cell Marker
Clone SPM310
Concentration 1mg/mL
Immunogen Pancreatic cancer-related mucin
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2a
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry (Paraffin)