missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-PINK1, Clone: 8E10.1D6, Novus Biologicals™

$139.00 - $329.00

Specifications

Antigen PINK1
Clone 8E10.1D6
Concentration 1.0mg/mL
Immunogen A synthetic peptide made to the human PINK1 protein sequence (between residues 100-250). [Swiss-Prot Q9BXM7]
Gene Accession No. Q9BXM7
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP236488 Novus Biologicals Canada
NBP236488
0.1mg Each for $329.00
NBP236488SS Novus Biologicals Canada
NBP236488SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

PINK1
1.0mg/mL
Q9BXM7
Protein G purified
Primary
Murine
Unlabeled
Purified
8E10.1D6
A synthetic peptide made to the human PINK1 protein sequence (between residues 100-250). [Swiss-Prot Q9BXM7]
65018
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2b, κ
Purified
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blot