Please login to your online account to display your discounted pricing

Mouse anti-Phospho-Tyrosine, Clone: SPM102, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen Phospho-Tyrosine
Clone SPM102
Concentration 0.2mg/mL
Immunogen Phosphotyrosine conjugated to KLH
Formulation PBS with 0.05% BSA.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23299001

 
novus biologicals canada
NBP23299001ML
0.1mL Each for $139.00

NBP23299010

 
novus biologicals canada
NBP23299010ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen Phospho-Tyrosine
Clone SPM102
Concentration 0.2mg/mL
Immunogen Phosphotyrosine conjugated to KLH
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity All
Isotype IgG2b
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting