Please login to your online account to display your discounted pricing

Mouse anti-Phospho-Tyrosine, Clone: SPM102, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23299010ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23299010

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity All
Antigen Phospho-Tyrosine
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Phosphotyrosine conjugated to KLH
Purification Method Purified
Quantity 1.0mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Clone SPM102
Concentration 0.2mg/mL
Conjugate Unlabeled
Isotype IgG2b
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified