Please login to your online account to display your discounted pricing

Mouse anti-Phospho-Tyrosine, Clone: SPM102, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23299001ML

 View all options for this product


For Research Use Only

Catalog No. NBP23299001


Description & Specifications


Antigen Phospho-Tyrosine
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM102
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity All
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Phosphotyrosine conjugated to KLH
Isotype IgG2b
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified