Please login to your online account to display your discounted pricing

Mouse anti-Phospho-Tyrosine, Clone: SPM102, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23473002MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23473002

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Host Species Mouse
Immunogen Phosphotyrosine conjugated to KLH
Quantity 0.2mg
Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 1mg/mL
Cross Reactivity All
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Conjugate Unlabeled
Reconstitution Protein A purified
Antigen Phospho-Tyrosine
Clone SPM102
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Isotype IgG2b
Test Specificity This MAb shows no cross-reaction with other phosphoamino acids and is superb for multiple applications including staining of formalin/paraffin tissues.
Purification Method Purified