Please login to your online account to display your discounted pricing

Mouse anti-Phospho-Tyrosine, Clone: SPM102, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23473001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23473001

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Phosphotyrosine conjugated to KLH
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Test Specificity This MAb shows no cross-reaction with other phosphoamino acids and is superb for multiple applications including staining of formalin/paraffin tissues.
Reconstitution Protein A purified
Conjugate Unlabeled
Antigen Phospho-Tyrosine
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Isotype IgG2b
Purification Method Purified
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Clone SPM102
Concentration 1mg/mL
Cross Reactivity All