Please login to your online account to display your discounted pricing

Mouse anti-PGP9.5 / UCHL-1, Clone: SPM575, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen PGP9.5 / UCHL-1
Clone SPM575
Concentration 0.2mg/mL
Immunogen Native UchL1 (PGP9.5) protein from brain
Gene Accession No. P09936
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23289601

 
novus biologicals canada
NBP23289601ML
0.1mL Each for $139.00

NBP23289610

 
novus biologicals canada
NBP23289610ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen PGP9.5 / UCHL-1
Clone SPM575
Concentration 0.2mg/mL
Immunogen Native UchL1 (PGP9.5) protein from brain
Gene Accession No. P09936
Gene ID (Entrez) 7345
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Cross Reactivity Zebrafish
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Western Blotting