Please login to your online account to display your discounted pricing

Mouse anti-PGP9.5 / UCHL-1, Clone: SPM575, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23289601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23289601

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Primary or Secondary Primary
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Cross Reactivity Zebrafish
Antigen PGP9.5 / UCHL-1
Format Purified
Gene Accession No. P09936
Host Species Mouse
Immunogen Native UchL1 (PGP9.5) protein from brain
Isotype IgG2a, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7345
Clone SPM575
Concentration 0.2mg/mL
Formulation PBS with 0.05% BSA.
Reconstitution Protein A purified