Please login to your online account to display your discounted pricing

Mouse anti-PGP9.5 / UCHL-1, Clone: SPM575, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23480802MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23480802

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Regulatory Status RUO
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Cross Reactivity Zebrafish
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Quantity 0.2mg
Purification Method Purified
Reconstitution Protein A purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene ID (Entrez) 7345
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity This MAb reacts with a protein of 20-30kDa, identified as PGP9.5, also known as ubiquitin carboxyl-terminal hydrolase-1 (UchL1).
Gene Accession No. P09936
Antigen PGP9.5 / UCHL-1
Concentration 1mg/mL
Primary or Secondary Primary
Format Purified
Conjugate Unlabeled
Isotype IgG2a, κ
Immunogen Native UchL1 (PGP9.5) protein from brain
Clone SPM575