Please login to your online account to display your discounted pricing

Mouse anti-PGP9.5 / UCHL-1, Clone: SPM575, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23480801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23480801

  • / Each
 

Description & Specifications

Specifications

Antigen PGP9.5 / UCHL-1
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Western Blot
Clone SPM575
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P09936
Host Species Murine
Immunogen Native UchL1 (PGP9.5) protein from brain
Isotype IgG2a, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7345
Test Specificity This MAb reacts with a protein of 20-30kDa, identified as PGP9.5, also known as ubiquitin carboxyl-terminal hydrolase-1 (UchL1).
Reconstitution Protein A purified
Target Species Bovine
Target Species Canine
Target Species Guinea Pig
Target Species Human
Target Species Murine
Target Species Porcine
Target Species Rabbit
Target Species Rat
Target Species Sheep
Target Species Zebrafish