Please login to your online account to display your discounted pricing

Mouse anti-PGP9.5 / UCHL-1, Clone: SPM574, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen PGP9.5 / UCHL-1
Clone SPM574
Concentration 0.2mg/mL
Immunogen Native UchL1 (PGP9.5) protein from brain
Gene Accession No. P09936
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23289501

 
novus biologicals canada
NBP23289501ML
0.1mL Each for $139.00

NBP23289510

 
novus biologicals canada
NBP23289510ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen PGP9.5 / UCHL-1
Clone SPM574
Concentration 0.2mg/mL
Immunogen Native UchL1 (PGP9.5) protein from brain
Gene Accession No. P09936
Gene ID (Entrez) 7345
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting