Please login to your online account to display your discounted pricing

Mouse anti-PGP9.5 / UCHL-1, Clone: SPM574, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23289501ML

 View all options for this product


For Research Use Only

Catalog No. NBP23289501


Description & Specifications


Antigen PGP9.5 / UCHL-1
Applications ELISA
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone SPM574
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P09936
Host Species Mouse
Immunogen Native UchL1 (PGP9.5) protein from brain
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7345
Reconstitution Protein A purified