missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-PGP9.5 / UCHL-1, Clone: SPM574, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen PGP9.5 / UCHL-1
Clone SPM574
Concentration 1mg/mL
Immunogen Native UchL1 (PGP9.5) protein from brain
Gene Accession No. P09936
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23480702 Novus Biologicals Canada
NBP23480702MG
0.2mg Each for $527.00
NBP23480701 Novus Biologicals Canada
NBP23480701MG
0.1mg Each for $384.00
Specifications Description & Specifications

Specifications

PGP9.5 / UCHL-1
1mg/mL
P09936
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM574
Native UchL1 (PGP9.5) protein from brain
7345
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Immunohistochemistry (Paraffin),Western Blot