Please login to your online account to display your discounted pricing

Mouse anti-PGP9.5 / UCHL-1, Clone: SPM574, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23480701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23480701

  • / Each
 

Description & Specifications

Specifications

Antigen PGP9.5 / UCHL-1
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P09936
Host Species Mouse
Applications ELISA
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Native UchL1 (PGP9.5) protein from brain
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7345
Test Specificity This MAb reacts with a protein of 20-30kDa, identified as PGP9.5, also known as ubiquitin carboxyl-terminal hydrolase-1 (UchL1).
Reconstitution Protein A purified
Clone SPM574
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat