Please login to your online account to display your discounted pricing

Mouse anti-PGP9.5 / UCHL-1, Clone: SPM574, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23480702MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23480702

  • / Each
 

Description & Specifications

Specifications

Antigen PGP9.5 / UCHL-1
Applications ELISA
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Clone SPM574
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P09936
Host Species Murine
Immunogen Native UchL1 (PGP9.5) protein from brain
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7345
Test Specificity This MAb reacts with a protein of 20-30kDa, identified as PGP9.5, also known as ubiquitin carboxyl-terminal hydrolase-1 (UchL1).
Reconstitution Protein A purified
Target Species Bovine
Target Species Human
Target Species Murine
Target Species Porcine
Target Species Rat