missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Perforin, Clone: BOR21, Novus Biologicals™

$274.00 - $329.00

Specifications

Antigen Perforin
Clone BOR21
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was Perforin.
Gene ID (Entrez) 5551
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14310601 Novus Biologicals Canada
NBP14310601MG
0.1mg Each for $329.00
NBP14310603 Novus Biologicals Canada
NBP1431060025MG
0.025mg Each for $274.00
Specifications Description & Specifications

Specifications

Perforin
0.2mg/mL
5551
Monoclonal
Store at 4°C. Do not freeze.
IgG2b
Affinity Purified
Western Blot
BOR21
The immunogen for this antibody was Perforin.
Phosphate buffer pH 7.2
Primary
Murine
Unlabeled
Affinity Purified