Please login to your online account to display your discounted pricing

Mouse anti-Perforin, Clone: BOR21, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1431060025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14310603

  • / Each
 

Description & Specifications

Specifications

Antigen Perforin
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Mouse
Immunogen The immunogen for this antibody was Perforin.
Cross Reactivity Human
Isotype IgG2b
Purification Method Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5551
Clone BOR21
Concentration 0.2mg/mL
Conjugate Unlabeled
Applications Western Blotting