Please login to your online account to display your discounted pricing

Mouse anti-PD-1, Clone: J116, Novus Biologicals™

$131.00 - $307.00

Specifications

Antigen PD-1
Clone J116
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was PD1.
Gene ID (Entrez) 5133
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14310703

 
novus biologicals canada
NBP1431070025MG
0.025mg Each for $131.00

NBP14310701

 
novus biologicals canada
NBP14310701MG
0.1mg Each for $307.00
Description & Specifications

Specifications

Antigen PD-1
Clone J116
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was PD1.
Gene ID (Entrez) 5133
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blot