Please login to your online account to display your discounted pricing

Mouse anti-PD-1, Clone: J116, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1431070025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14310703

  • / Each
 

Description & Specifications

Specifications

Antigen PD-1
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blot
Clone J116
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Murine
Immunogen The immunogen for this antibody was PD1.
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5133
Target Species Human