Please login to your online account to display your discounted pricing

Mouse anti-PCNA, Clone: SPM350, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen PCNA
Clone SPM350
Concentration 0.2mg/mL
Immunogen Rat PCNA/Protein A fusion protein
Gene Accession No. P12004
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23298901

 
novus biologicals canada
NBP23298901ML
0.1mL Each for $139.00

NBP23298910

 
novus biologicals canada
NBP23298910ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen PCNA
Clone SPM350
Concentration 0.2mg/mL
Immunogen Rat PCNA/Protein A fusion protein
Gene Accession No. P12004
Gene ID (Entrez) 5111
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Chicken
Cross Reactivity Drosophila
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Cross Reactivity Yeast
Cross Reactivity Zebrafish
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting