Please login to your online account to display your discounted pricing

Mouse anti-PCNA, Clone: SPM350, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23298910ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23298910

  • / Each
 

Description & Specifications

Specifications

Antigen PCNA
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM350
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Drosophila
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Cross Reactivity Yeast
Cross Reactivity Zebrafish
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P12004
Host Species Mouse
Immunogen Rat PCNA/Protein A fusion protein
Isotype IgG2a, κ
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5111
Test Specificity Drosophila melanogaster, Yeast (S pombe, S cerevisiae)
Reconstitution Protein A purified