Please login to your online account to display your discounted pricing

Mouse anti-PCNA, Clone: SPM350, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23476701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23476701

  • / Each
 

Description & Specifications

Specifications

Immunogen Rat PCNA/Protein A fusion protein
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Antigen PCNA
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P12004
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG2a, κ
Purification Method Purified
Quantity 0.1mg
Primary or Secondary Primary
Test Specificity Recognizes a non-histone protein of 36kDa, which is identified as proliferating cell nuclear antigen (PCNA). It is also known as cyclin or polymerase delta auxiliary protein. Elevated expression of PCNA/cyclin has been shown in the nucleus during late G1 phase immediately before the onset of DNA synthesis, becoming maximal during S-phase and declining during G2 and M phases. This MAb is excellent for multiple applications. Reacts with Yeast (S. pombe, S. cerevisiae).
Clone SPM350
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Drosophila
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Cross Reactivity Yeast
Cross Reactivity Zebrafish
Regulatory Status RUO
Gene ID (Entrez) 5111
Reconstitution Protein A purified