Please login to your online account to display your discounted pricing

Mouse anti-PCNA, Clone: PC10, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen PCNA
Clone PC10
Concentration 1mg/mL
Immunogen Rat PCNA/Protein A fusion protein
Gene Accession No. P12004
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23470701

 
novus biologicals canada
NBP23470701MG
0.1mg Each for $384.00

NBP23470702

 
novus biologicals canada
NBP23470702MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen PCNA
Clone PC10
Concentration 1mg/mL
Immunogen Rat PCNA/Protein A fusion protein
Gene Accession No. P12004
Gene ID (Entrez) 5111
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Chicken
Cross Reactivity Drosophila
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Cross Reactivity Yeast
Cross Reactivity Zebrafish
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting