Please login to your online account to display your discounted pricing

Mouse anti-PCNA, Clone: PC10, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23470702MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23470702

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity Drosophila melanogaster, Yeast (S pombe S cerevisiae)
Regulatory Status RUO
Isotype IgG2a, κ
Gene ID (Entrez) 5111
Cross Reactivity Chicken
Cross Reactivity Drosophila
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Cross Reactivity Yeast
Cross Reactivity Zebrafish
Conjugate Unlabeled
Quantity 0.2mg
Immunogen Rat PCNA/Protein A fusion protein
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Clone PC10
Concentration 1mg/mL
Gene Accession No. P12004
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen PCNA
Format Purified
Purification Method Purified
Reconstitution Protein A purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.