Please login to your online account to display your discounted pricing

Mouse anti-PCNA, Clone: PC10, Azide Free, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23470701MG

 View all options for this product


For Research Use Only

Catalog No. NBP23470701


Description & Specifications


Antigen PCNA
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone PC10
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Drosophila
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Cross Reactivity Yeast
Cross Reactivity Zebrafish
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P12004
Host Species Mouse
Immunogen Rat PCNA/Protein A fusion protein
Isotype IgG2a, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5111
Test Specificity Drosophila melanogaster, Yeast (S pombe S cerevisiae)
Reconstitution Protein A purified