Please login to your online account to display your discounted pricing

Mouse anti-Pax7, Clone: PAX7/497, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23470601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23470601

  • / Each
 

Description & Specifications

Specifications

Antigen Pax7
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blot
Clone PAX7/497
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P23759
Host Species Murine
Immunogen Recombinant human PAX7 protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5081
Reconstitution Protein A purified
Target Species Chicken
Target Species Human
Target Species Murine
Target Species Rat
Target Species Zebrafish