Please login to your online account to display your discounted pricing

Mouse anti-Pax6, Clone: PAX6/496, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Pax6
Clone PAX6/496
Concentration 1mg/mL
Immunogen Recombinant human PAX6 protein
Gene Accession No. P26367
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23470502

 
novus biologicals canada
NBP23470502MG
0.2mg Each for $527.00

NBP23470501

 
novus biologicals canada
NBP23470501MG
0.1mg Each for $384.00
Description & Specifications

Specifications

Antigen Pax6
Clone PAX6/496
Concentration 1mg/mL
Immunogen Recombinant human PAX6 protein
Gene Accession No. P26367
Gene ID (Entrez) 5080
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blot