Please login to your online account to display your discounted pricing

Mouse anti-Pax6, Clone: PAX6/496, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23470502MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23470502

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P26367
Host Species Mouse
Isotype IgG1, κ
Quantity 0.2mg
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone PAX6/496
Concentration 1mg/mL
Antigen Pax6
Immunogen Recombinant human PAX6 protein
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 5080
Reconstitution Protein A purified
Conjugate Unlabeled