Please login to your online account to display your discounted pricing

Mouse anti-Pax5/BSAP, Clone: 7D3, Novus Biologicals™

$367.12 - $367.12

Specifications

Antigen Pax5/BSAP
Clone 7D3
Concentration 1mg/mL
Immunogen Purified recombinant fragment of human PAX5 (AA: 235-382) expressed in E. coli.
Gene ID (Entrez) 5079
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP237464

 
novus biologicals canada
NBP237464
0.1mL Each for $367.12
Description & Specifications

Specifications

Antigen Pax5/BSAP
Clone 7D3
Concentration 1mg/mL
Immunogen Purified recombinant fragment of human PAX5 (AA: 235-382) expressed in E. coli.
Gene ID (Entrez) 5079
Formulation PBS
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications ELISA
Applications Immunohistochemistry
Applications Western Blotting