Please login to your online account to display your discounted pricing

Mouse anti-Pax5/BSAP, Clone: 7D3, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP237478

 View all options for this product


For Research Use Only

Catalog No. NBP237478


Description & Specifications


Antigen Pax5/BSAP
Applications ELISA
Applications Western Blotting
Clone 7D3
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation PBS
Host Species Mouse
Immunogen Purified recombinant fragment of human PAX5 (AA: 235-382) expressed in E. coli.
Isotype IgG1
Purification Method Affinity Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5079