Please login to your online account to display your discounted pricing

Mouse anti-pan Cytokeratin, Clone: SPM583, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23438605ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23438605

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Keratin-enriched preparation from cultured human A431
Purification Method Purified
Quantity 0.5mL
Primary or Secondary Primary
Gene ID (Entrez) 3848
Test Specificity This MAb recognizes cytokeratin 4, 5, 6, 8, 10, 13, and 18. Reacts with Frog, Marmoset.
Clone SPM583
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Amphibian
Cross Reactivity Bovine
Cross Reactivity Goat
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Antigen pan Cytokeratin
Formulation PBS with 0.05% BSA. Protein G purified.
Isotype IgG1
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal