Please login to your online account to display your discounted pricing

Mouse anti-pan Cytokeratin, Clone: SPM583, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23438601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23438601

  • / Each
 

Description & Specifications

Specifications

Antigen pan Cytokeratin
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone SPM583
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Amphibian
Cross Reactivity Bovine
Cross Reactivity Goat
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA. Protein G purified.
Host Species Mouse
Immunogen Keratin-enriched preparation from cultured human A431
Isotype IgG1
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3848
Test Specificity This MAb recognizes cytokeratin 4, 5, 6, 8, 10, 13, and 18. Reacts with Frog, Marmoset.