missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-pan Cytokeratin, Clone: SPM583, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen pan Cytokeratin
Clone SPM583
Concentration 1mg/mL
Immunogen Keratin-enriched preparation from cultured human A431
Gene ID (Entrez) 3848
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23443301 Novus Biologicals Canada
NBP23443301MG
0.1mg Each for $384.00
NBP23443302 Novus Biologicals Canada
NBP23443302MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

pan Cytokeratin
1mg/mL
3848
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Purified
Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Western Blot
SPM583
Keratin-enriched preparation from cultured human A431
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified