Please login to your online account to display your discounted pricing

Mouse anti-pan Cytokeratin, Clone: SPM583, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23443301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23443301

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Amphibian
Cross Reactivity Bovine
Cross Reactivity Goat
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Primary or Secondary Primary
Immunogen Keratin-enriched preparation from cultured human A431
Gene ID (Entrez) 3848
Monoclonal or Polyclonal Monoclonal
Quantity 0.1mg
Regulatory Status RUO
Reconstitution Protein A purified
Conjugate Unlabeled
Host Species Mouse
Test Specificity This MAb recognizes cytokeratin 4, 5, 6, 8, 10, 13, and 18. Reacts with Frog, Marmoset.
Antigen pan Cytokeratin
Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Clone SPM583
Purification Method Purified
Concentration 1mg/mL