Please login to your online account to display your discounted pricing

Mouse anti-pan Cytokeratin, Clone: C11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23433405ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23433405

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Amphibian
Cross Reactivity Bovine
Cross Reactivity Frog
Cross Reactivity Goat
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Marmoset
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Conjugate Unlabeled
Quantity 0.5mL
Gene Accession No. P19013
Host Species Mouse
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Antigen pan Cytokeratin
Clone C11
Immunogen Keratin-enriched preparation from cultured human A431
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3848
Isotype IgG1
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Format Purified
Concentration 0.2mg/mL