Please login to your online account to display your discounted pricing

Mouse anti-pan Cytokeratin, Clone: C11, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23433401ML

 View all options for this product


For Research Use Only

Catalog No. NBP23433401


Description & Specifications


Antigen pan Cytokeratin
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone C11
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Amphibian
Cross Reactivity Bovine
Cross Reactivity Frog
Cross Reactivity Goat
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Marmoset
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA. Protein G purified.
Gene Accession No. P19013
Host Species Mouse
Immunogen Keratin-enriched preparation from cultured human A431
Isotype IgG1
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3848