Please login to your online account to display your discounted pricing

Mouse anti-pan Cytokeratin, Clone: C11, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen pan Cytokeratin
Clone C11
Concentration 1mg/mL
Immunogen Keratin-enriched preparation from cultured human A431
Gene Accession No. P19013
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23457201

 
novus biologicals canada
NBP23457201MG
0.1mg Each for $384.00

NBP23457202

 
novus biologicals canada
NBP23457202MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen pan Cytokeratin
Clone C11
Concentration 1mg/mL
Immunogen Keratin-enriched preparation from cultured human A431
Gene Accession No. P19013
Gene ID (Entrez) 3848
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Amphibian
Cross Reactivity Bovine
Cross Reactivity Frog
Cross Reactivity Goat
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Marmoset
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting