Please login to your online account to display your discounted pricing

Mouse anti-pan Cytokeratin, Clone: C11, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23457201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23457201

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Amphibian
Cross Reactivity Bovine
Cross Reactivity Frog
Cross Reactivity Goat
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Marmoset
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Antigen pan Cytokeratin
Regulatory Status RUO
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Reconstitution Protein A purified
Clone C11
Concentration 1mg/mL
Conjugate Unlabeled
Primary or Secondary Primary
Immunogen Keratin-enriched preparation from cultured human A431
Format Purified
Host Species Mouse
Purification Method Purified
Isotype IgG1
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Accession No. P19013
Quantity 0.1mg
Gene ID (Entrez) 3848