Please login to your online account to display your discounted pricing

Mouse anti-Pan Beta Crystallins, Clone: 3.H9.2, Novus Biologicals™

$309.05 - $518.93

Specifications

Antigen Pan Beta Crystallins
Clone 3.H9.2
Immunogen Native bovine beta-crystallin.
Formulation Liquid. In PBS containing 50% glycerol. Protein G purified.
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP19787005

 
novus biologicals canada
NBP197870005MG
0.05mg Each for $309.05

NBP19787002

 
novus biologicals canada
NBP19787002MG
0.2mg Each for $518.93
Description & Specifications

Specifications

Antigen Pan Beta Crystallins
Clone 3.H9.2
Immunogen Native bovine beta-crystallin.
Formulation Liquid. In PBS containing 50% glycerol. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Western Blot