Please login to your online account to display your discounted pricing

Mouse anti-Pan Beta Crystallins, Clone: 3.H9.2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP19787002MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP19787002

  • / Each
 

Description & Specifications

Specifications

Applications Western Blotting
Antigen Pan Beta Crystallins
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Conjugate Unlabeled
Format Purified
Host Species Mouse
Immunogen Native bovine beta-crystallin.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Bovine
Cross Reactivity Rabbit
Formulation Liquid. In PBS containing 50% glycerol. Protein G purified.
Isotype IgG1
Purification Method Purified
Clone 3.H9.2