Please login to your online account to display your discounted pricing

Mouse anti-Pan Beta Crystallins, Clone: 3.H9.2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP197870005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP19787005

  • / Each
 

Description & Specifications

Specifications

Antigen Pan Beta Crystallins
Applications Western Blot
Clone 3.H9.2
Conjugate Unlabeled
Format Purified
Formulation Liquid. In PBS containing 50% glycerol. Protein G purified.
Host Species Murine
Immunogen Native bovine beta-crystallin.
Isotype IgG1
Purification Method Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Bovine
Target Species Rabbit