Please login to your online account to display your discounted pricing

Mouse anti-pan Actin, Clone: SPM160, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen pan Actin
Clone SPM160
Concentration 0.2mg/mL
Immunogen SDS extract of human myocardium.
Gene ID (Entrez) 58
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23435101

 
novus biologicals canada
NBP23435101ML
0.1mL Each for $139.00

NBP23435105

 
novus biologicals canada
NBP23435105ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen pan Actin
Clone SPM160
Concentration 0.2mg/mL
Immunogen SDS extract of human myocardium.
Gene ID (Entrez) 58
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Rabbit
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting