Please login to your online account to display your discounted pricing

Mouse anti-pan Actin, Clone: SPM160, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23435105ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23435105

  • / Each
 

Description & Specifications

Specifications

Antigen pan Actin
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen SDS extract of human myocardium.
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 58
Test Specificity This antibody recognizes actin of skeletal, cardiac, and smooth muscle cells. It is not reactive with other mesenchymal cells except for myoepithelium.
Reconstitution Protein A purified
Clone SPM160
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rabbit
Cross Reactivity Rat
Concentration 0.2mg/mL