Please login to your online account to display your discounted pricing

Mouse anti-pan Actin, Clone: SPM160, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen pan Actin
Clone SPM160
Concentration 1mg/mL
Immunogen SDS extract of human myocardium.
Gene ID (Entrez) 58
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23439901

 
novus biologicals canada
NBP23439901MG
0.1mg Each for $384.00

NBP23439902

 
novus biologicals canada
NBP23439902MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen pan Actin
Clone SPM160
Concentration 1mg/mL
Immunogen SDS extract of human myocardium.
Gene ID (Entrez) 58
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot