Please login to your online account to display your discounted pricing

Mouse anti-pan Actin, Clone: SPM160, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23439901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23439901

  • / Each
 

Description & Specifications

Specifications

Antigen pan Actin
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone SPM160
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Murine
Immunogen SDS extract of human myocardium.
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 58
Test Specificity This antibody recognizes actin of skeletal, cardiac, and smooth muscle cells. It is not reactive with other mesenchymal cells except for myoepithelium.
Reconstitution Protein A purified
Target Species Human
Target Species Rabbit
Target Species Rat